Learn More
Medchemexpress LLC Ethyl stearate | 111-61-5 | 98.8% | 312.54 | 1 G
SDP
Catalog No. 5000454549
Encompass_Preferred
Ethyl stearate is an ester compound produced by the reaction of ethanol and stearic acid. It is commonly used as a flavoring agent in various foods such as baked goods, beverages and desserts. Ethyl stearate is also used as a fragrance ingredient in the manufacture of perfume, cologne and personal care products. Additionally, it has applications in industrial settings such as the production of lubricants and plasticizers. This product is a biochemical reagent suitable for life science related research.
  • Used as a flavoring agent in foods
  • Fragrance ingredient in personal care products
  • Applied in lubricants and plasticizers production
  • Suitable as a biochemical reagent for life science research
Catalog No. 50-004-54549 Supplier Medchemexpress LLC Supplier No. HYW0138091G
Ethyl stearate is an ester compound produced by the reaction of ethanol and stearic acid. It is commonly used as a flavoring agent in various foods such as baked goods, beverages and desserts. Ethyl stearate is also used as a fragrance ingredient in the manufacture of perfume, cologne and personal care products. Additionally, it has applications in industrial settings such as the production of lubricants and plasticizers. This product is a biochemical reagent suitable for life science related research.
  • Used as a flavoring agent in foods
  • Fragrance ingredient in personal care products
  • Applied in lubricants and plasticizers production
  • Suitable as a biochemical reagent for life science research