Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl stearate | 111-61-5 | 98.8% | 312.54 | 50 MG
SDP

Catalog No. 5000454556
Encompass_Preferred

Ethyl stearate is an ester compound formed from ethanol and stearic acid. It serves as a versatile ingredient, commonly employed as a flavoring agent in various foods, a fragrance component in personal care products, and in industrial applications such as lubricants and plasticizers. Additionally, it functions as a biochemical reagent for life science research.

  • Used as a flavoring agent in foods, beverages, and desserts.
  • Functions as a fragrance ingredient in perfumes, colognes, and personal care products.
  • Has industrial applications in lubricants and plasticizers.
  • Serves as a biochemical reagent for life science research.
  • Purity of 98.82%.
  • Molecular weight of 312.54.
  • Formulated as C20H40O2.
  • Appears as a solid below 34°C and a liquid above 38°C.
  • Soluble in DMSO.
  • Stable for 3 years at -20°C in pure form.

Catalog No. 50-004-54556 Supplier Medchemexpress LLC Supplier No. HYW01380950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl stearate is an ester compound formed from ethanol and stearic acid. It serves as a versatile ingredient, commonly employed as a flavoring agent in various foods, a fragrance component in personal care products, and in industrial applications such as lubricants and plasticizers. Additionally, it functions as a biochemical reagent for life science research.

  • Used as a flavoring agent in foods, beverages, and desserts.
  • Functions as a fragrance ingredient in perfumes, colognes, and personal care products.
  • Has industrial applications in lubricants and plasticizers.
  • Serves as a biochemical reagent for life science research.
  • Purity of 98.82%.
  • Molecular weight of 312.54.
  • Formulated as C20H40O2.
  • Appears as a solid below 34°C and a liquid above 38°C.
  • Soluble in DMSO.
  • Stable for 3 years at -20°C in pure form.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.