Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Sulfide 98%, Thermo Scientific™

Catalog No. AC148095000
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC148095000 500 mL
AC148090100 10 mL
AC148091000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC148095000 Supplier Thermo Scientific Chemicals Supplier No. 148095000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-93-2
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.18
MDL Number MFCD00009270
InChI Key LJSQFQKUNVCTIA-UHFFFAOYSA-N
Synonym diethyl sulfide, ethyl sulfide, ethyl thioether, 3-thiapentane, diethylthioether, thioethyl ether, ethylthioethane, 1,1'-thiobisethane, diethyl sulphide, ethane, 1,1'-thiobis
PubChem CID 9609
ChEBI CHEBI:27710
IUPAC Name (ethylsulfanyl)ethane
SMILES CCSCC

Specifications

Color Colorless
Density 0.83
Infrared Spectrum Authentic
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Linear Formula (C2H5)2S
Quantity 500 mL
Beilstein 01, 344
Merck Index 15, 3905
Formula Weight 90.18
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Ethyl sulfide, 98%
Show More Show Less
Hazard Category STENCH
EINECSNumber 206-526-9
RTECSNumber LC7200000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.