Learn More
Medchemexpress LLC Ethyl tetradecanoate 500g
SDP
Catalog No. 5000206159
Encompass_Preferred
Ethyl tetradecanoate which is an ester formed by the condensation of ethanol and myristic acid is commonly used as a flavoring agent and fragrance in the food and cosmetic industries
Catalog No. 50-002-06159 Supplier Medchemexpress LLC Supplier No. HYW099586500G
Ethyl tetradecanoate which is an ester formed by the condensation of ethanol and myristic acid is commonly used as a flavoring agent and fragrance in the food and cosmetic industries