Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl trans-2-decenoate | 7367-88-6 | MFCD00015299 | 97.0% | 198.30 g·mol⁻¹ | C12H22O2 | 5 G
SDP

Catalog No. 5000282597
Encompass_Preferred

Ethyl trans-2-decenoate is the ethyl ester of 2-decenoic acid with a fruity, floral aroma used as a flavoring and fragrance reagent and as a reference standard in research and formulation.

  • High purity (97.0%).
  • Colorless to light yellow liquid form.
  • Molecular formula C12H22O2; molecular weight 198.30 g·mol⁻¹.
  • Available in laboratory sizes: 5 G, 10 G, 25 G, 50 G.
  • COA and SDS available for batch-specific data and handling guidance.

Catalog No. 50-002-82597 Supplier Medchemexpress LLC Supplier No. HYW1275735G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl trans-2-decenoate is the ethyl ester of 2-decenoic acid with a fruity, floral aroma used as a flavoring and fragrance reagent and as a reference standard in research and formulation.

  • High purity (97.0%).
  • Colorless to light yellow liquid form.
  • Molecular formula C12H22O2; molecular weight 198.30 g·mol⁻¹.
  • Available in laboratory sizes: 5 G, 10 G, 25 G, 50 G.
  • COA and SDS available for batch-specific data and handling guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.