Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl trans-2-Hexenoate 97.0+%, TCI America™
SDP

Catalog No. E078725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E078725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E078725G Supplier TCI America Supplier No. E078725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27829-72-7
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00036541
InChI Key SJRXWMQZUAOMRJ-VOTSOKGWSA-N
Synonym ethyl hex-2-enoate, ethyl 2-hexenoate, ethyl trans-2-hexenoate, 2-hexenoic acid, ethyl ester, ethyl e-hex-2-enoate, ethyl e-2-hexenoate, ethyl 2e-hexenoate, 2-hexenoic acid, ethyl ester, 2e, unii-ol4euz6c13, 2-hexenoic acid, ethyl ester, e
PubChem CID 5364778
IUPAC Name ethyl (E)-hex-2-enoate
SMILES CCCC=CC(=O)OCC

Specifications

Color Colorless
Boiling Point 85°C
Quantity 25 g
Formula Weight 142.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl trans-2-Hexenoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.