Learn More
Medchemexpress LLC Ethyl trans-4-decenoate | 76649-16-6 | MFCD00015574 | >98.0% | 198.30 | C12H22O2 | 25 G
SDP
Catalog No. 5000283528
Encompass_Preferred
Ethyl trans-4-decenoate is the ethyl ester of trans-4-decenoic acid used as an organic reagent in biochemical assays and research. It typically appears as a colorless liquid with formula C12H22O2 and molecular weight 198.30.
  • Useful as a biochemical assay reagent and organic synthesis intermediate.
  • Identified by CAS number 76649-16-6 for unambiguous chemical reference.
  • Liquid, colorless to pale yellow appearance suitable for formulation.
  • Stable under recommended storage; in solvent: -80°C for 2 years, -20°C for 1 year.
  • Characterized by molecular formula C12H22O2 and molecular weight 198.30.
Catalog No. 50-002-83528 Supplier Medchemexpress LLC Supplier No. HYW12758325G
Ethyl trans-4-decenoate is the ethyl ester of trans-4-decenoic acid used as an organic reagent in biochemical assays and research. It typically appears as a colorless liquid with formula C12H22O2 and molecular weight 198.30.
  • Useful as a biochemical assay reagent and organic synthesis intermediate.
  • Identified by CAS number 76649-16-6 for unambiguous chemical reference.
  • Liquid, colorless to pale yellow appearance suitable for formulation.
  • Stable under recommended storage; in solvent: -80°C for 2 years, -20°C for 1 year.
  • Characterized by molecular formula C12H22O2 and molecular weight 198.30.