Learn More
Medchemexpress LLC Ethyl trans-cinnamate-d5 | 856765-68-9 | MFCD12022206 | 99.7% | 181.24 | C11H7D5O2 | 50mg
SDP
Catalog No. 5000269862
Encompass_Preferred
Ethyl cinnamate-d5 is the deuterium labeled Ethyl cinnamate[1] Ethyl cinnamate is a fragrance ingredient used in many fragrance compounds Ethyl cinnamate is a food flavor and additive for cosmetic products Ethyl cinnamate is also an excellent clearing reagent for mammalian tissues[2][3]
Catalog No. 50-002-69862 Supplier Medchemexpress LLC Supplier No. HYY0121S50MG
Ethyl cinnamate-d5 is the deuterium labeled Ethyl cinnamate[1] Ethyl cinnamate is a fragrance ingredient used in many fragrance compounds Ethyl cinnamate is a food flavor and additive for cosmetic products Ethyl cinnamate is also an excellent clearing reagent for mammalian tissues[2][3]