Learn More
Medchemexpress LLC Ethyl Tridecanoate 25g
SDP
Catalog No. 5000206846
Encompass_Preferred
Ethyl Tridecanoate is an ester compound with a fruity odor commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of cosmetics personal care products and cleaning agents
Catalog No. 50-002-06846 Supplier Medchemexpress LLC Supplier No. 25GHYW127439
Ethyl Tridecanoate is an ester compound with a fruity odor commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of cosmetics personal care products and cleaning agents