Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl trifluoroacetate

Catalog No. E50000100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E50000100G 100 g
E5000025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E50000100G Supplier Sigma Aldrich Supplier No. E50000100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl trifluoroacetate can be used:
  • To synthesize cyclopentenones or furans containing trifluoromethyl group from aromatic ynones
  • In the selective trifluoroacetylation of anilines catalyzed by 4-dimethylaminopyridine
  • As a starting material in the two-step electrosynthesis of trifluoroacetyltrimethylsilane (cf3cosime3)
  • In the preparation of trifluoromethyl ketones via trifluoroacetic ester/ketone metathesis with alkyl aryl ketones
  • To synthesize o-fluorinated trifluoro acetophenones from substituted fluorobenzene

Specifications

CAS 383-63-1
Density 1.194 g/mL (at 25°C (literature))
Boiling Point 60°C to 62°C (lit.)
Molecular Formula C4H5F3O2
Refractive Index n20/D 1.307 (literature)
Linear Formula CF3COOC2H5
MDL Number MFCD00000419
Quantity 100 g
Synonym Trifluoroacetic acid ethyl ester
Molecular Weight (g/mol) 142.08
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.