Learn More
Dichloromethane, for residue and pestic. anal., stab. with amylene
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.
Specifications
Specifications
| Melting Point | -97.0°C |
| Decomposition Information | >120°C |
| Density | 1.3250g/mL |
| Quantity | 2.5 L |
| Physical Form | Liquid |
| Residue after Evaporation | 2ppm max. |
| Specific Gravity | 1.325 |
| Vapor Pressure | Vapor Pressure: 475hPa at 20°C |
| Linear Formula | CH2Cl2 |
| Viscosity | 0.43 mPa.s (20°C) |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician. |
| EINECSNumber | 200-838-9 |
| RTECSNumber | PA8050000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.