Learn More
Medchemexpress LLC Ethyl (triphenylphosphoranylidene) acetate | 1099-45-2 | 99.43% | C22H21O2P | 100 G
SDP
Catalog No. 5000433899
Encompass_Preferred
Ethyl (triphenylphosphoranylidene) acetate, also known as (Carbethoxymethylene)triphenylphosphorane, is a biochemical reagent suitable for life science research. It may act as an inhibitor of cholinesterase, affecting AChE and BChE.
  • Biochemical reagent for life science research
  • Can be used as a biological material or organic compound
  • May act as a cholinesterase inhibitor
  • Inhibits AChE and BChE
Catalog No. 50-004-33899 Supplier Medchemexpress LLC Supplier No. HY76446100G
Ethyl (triphenylphosphoranylidene) acetate, also known as (Carbethoxymethylene)triphenylphosphorane, is a biochemical reagent suitable for life science research. It may act as an inhibitor of cholinesterase, affecting AChE and BChE.
  • Biochemical reagent for life science research
  • Can be used as a biological material or organic compound
  • May act as a cholinesterase inhibitor
  • Inhibits AChE and BChE