Learn More
Medchemexpress LLC Ethyl undecanoate | 627-90-7 | MFCD00008958 | 99.7% | 214.35 g·mol⁻¹ | C13H26O2 | 100 G
SDP
Catalog No. 5000302774
Encompass_Preferred
Ethyl undecanoate is the ethyl ester of undecanoic acid supplied as a high-purity organic reagent for research, analytical standards, and formulation work in flavor and fragrance development.
  • High purity suitable for analytical and research use.
  • Applicable as a flavor and fragrance ingredient in formulation studies.
  • Suitable as a reference standard for GC/MS and other analytical methods.
  • Liquid organic ester with formula C13H26O2 and molecular weight 214.35 g·mol⁻¹.
Catalog No. 50-003-02774 Supplier Medchemexpress LLC Supplier No. HYW014207100G
Ethyl undecanoate is the ethyl ester of undecanoic acid supplied as a high-purity organic reagent for research, analytical standards, and formulation work in flavor and fragrance development.
  • High purity suitable for analytical and research use.
  • Applicable as a flavor and fragrance ingredient in formulation studies.
  • Suitable as a reference standard for GC/MS and other analytical methods.
  • Liquid organic ester with formula C13H26O2 and molecular weight 214.35 g·mol⁻¹.