Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl vanillate | 617-05-0 | 99.69% | 100 G
SDP

Catalog No. 5000413166
Encompass_Preferred

Ethyl Vanillate is an off-white to light yellow solid compound. It functions as a fungicidal agent, inhibiting 17β-HSD2 with an IC50 of 1.3 μM. This product is intended for research use only.

  • Fungicidal agent
  • Inhibits 17β-HSD2
  • IC50 of 1.3 μM
  • Molecular weight: 196.20
  • Formula: C10H12O4
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Shipping: Room temperature
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-13166 Supplier Medchemexpress LLC Supplier No. HYB1643100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl Vanillate is an off-white to light yellow solid compound. It functions as a fungicidal agent, inhibiting 17β-HSD2 with an IC50 of 1.3 μM. This product is intended for research use only.

  • Fungicidal agent
  • Inhibits 17β-HSD2
  • IC50 of 1.3 μM
  • Molecular weight: 196.20
  • Formula: C10H12O4
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Shipping: Room temperature
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.