Learn More
Selleck Chemical LLC Ethyl Vanillate S3993-25mg
SDP
Catalog No. 503109546
Encompass_Preferred
Ethyl vanillin (Bourbonal Ethylprotal) a vanillin analog is a flavouring agent and a fungicidal agent that exhibits strong antioxidant activity Ethyl Vanillate inhibits 17 -HSD2 with an IC50 1 3 M
Catalog No. 50-310-9546 Supplier Selleck Chemical LLC Supplier No. S399325MG
Ethyl vanillin (Bourbonal Ethylprotal) a vanillin analog is a flavouring agent and a fungicidal agent that exhibits strong antioxidant activity Ethyl Vanillate inhibits 17 -HSD2 with an IC50 1 3 M