Learn More
Medchemexpress LLC Ethyl vanillin acetate | 72207-94-4 | MFCD01666413 | 99.7% | 208.21 g/mol | C11H12O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10782010MM 1ML
Encompass_Preferred
Ethyl vanillin acetate is the acetate ester of ethyl vanillin with a vanilla-like aroma, supplied for laboratory research use. The compound is offered as a 10 mM solution in DMSO and as a solid powder. Typical applications include flavor and fragrance research, sensory studies, and use as a synthetic intermediate in organic chemistry.
  • Vanilla-like aroma profile.
  • Available as 10 mM solution in DMSO and as solid powder.
  • High purity suitable for analytical and research applications.
  • Useful in flavor, fragrance, and sensory research.
  • CAS 72207-94-4 for unambiguous identification.
Catalog No. 50-002-44518