Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Vanillin, NF, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18602549 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
40 kg
Packaging:
Amber Glass Bottle
Drum
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-549 125 g Amber Glass Bottle
18-602-550 500 g Amber Glass Bottle
18-602-551 2.5 kg Poly Pail
18-602-552 12 kg Fiber Drum
18-602-553 40 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-549 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ET119125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

ET119, 121-32-4, C9H10O3

Spectrum™ Chemical Ethyl Vanillin, NF is used as a flavorant. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 121-32-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00006944
InChI Key CBOQJANXLMLOSS-UHFFFAOYSA-N
IUPAC Name 3-ethoxy-4-hydroxybenzaldehyde
SMILES CCOC1=CC(C=O)=CC=C1O

Specifications

CAS Max % 100%
Loss on Drying 1%
Melting Range 76-78°C
Packaging Amber Glass Bottle
Quantity 125 g
Percent Purity 98 to 101%
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.