Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethylaluminum Dichloride Solution

Catalog No. 251615800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-161-5800ML 800 mL
25-161-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 25-161-5800ML Supplier Sigma Aldrich Supplier No. 251615800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethylaluminum dichloride is a strong Lewis acid and a proton scavenger that can be used:To catalyze polymerization of isobutylene.To facilitate Diels-Alder reaction of +a,+a-unsaturated esters.To carry out Pinacol reduction rearrangements.To promote Schmidt rearrangement of diketoazides in the synthesis of indolizidines and pyrroloazepinediones.

Specifications

CAS 563-43-9
Density 0.729 g/mL at 25°C
Molecular Formula C2H5AlCl2
Linear Formula CH3CH2AlCl2
Quantity 800 mL
MDL Number MFCD00000457
Synonym Dichloro(ethyl)alumane solution
Molecular Weight (g/mol) 126.95 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.