Learn More
Medchemexpress LLC Ethylbenzene-2,3,4,5,6-d5 | 20302-26-5 | C8H5D5 | 25 MG
SDP
Catalog No. 5000374501
Encompass_Preferred
Ethylbenzene-2,3,4,5,6-d5 is a deuterium-labeled form of Ethylbenzene, intended for research use only. This compound is part of a class of stable isotopes where hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily to serve as tracers for quantitative analysis during drug development processes. Deuteration has attracted attention due to its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-74501 Supplier Medchemexpress LLC Supplier No. 25MGHY150844S
Ethylbenzene-2,3,4,5,6-d5 is a deuterium-labeled form of Ethylbenzene, intended for research use only. This compound is part of a class of stable isotopes where hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily to serve as tracers for quantitative analysis during drug development processes. Deuteration has attracted attention due to its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.