Learn More
Medchemexpress LLC Ethylbenzene-2,3,4,5,6-d5 | 20302-26-5 | MFCD00039860 | C8H5D5 | 50 MG
SDP
Catalog No. 5000374990
Encompass_Preferred
Ethylbenzene-2,3,4,5,6-d5 is a deuterium-labeled Ethylbenzene, used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs. This compound is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-74990 Supplier Medchemexpress LLC Supplier No. HY150844S50MG
Ethylbenzene-2,3,4,5,6-d5 is a deuterium-labeled Ethylbenzene, used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs. This compound is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS