Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethylbenzene-2,3,4,5,6-d5 | 20302-26-5 | MFCD00064518 | C8H5D5 | 100 MG
SDP

Catalog No. 5000375726
Encompass_Preferred

Ethylbenzene-2,3,4,5,6-d5 is a deuterium labeled Ethylbenzene. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.

  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • Affects pharmacokinetic and metabolic profiles of drugs
  • Purity: 99.25%
  • Molecular weight: 111.20
  • Appearance: Liquid
  • Color: Colorless to light yellow

Catalog No. 50-003-75726 Supplier Medchemexpress LLC Supplier No. HY150844S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethylbenzene-2,3,4,5,6-d5 is a deuterium labeled Ethylbenzene. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.

  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • Affects pharmacokinetic and metabolic profiles of drugs
  • Purity: 99.25%
  • Molecular weight: 111.20
  • Appearance: Liquid
  • Color: Colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.