Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylbenzene, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019822 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9822 5 mL
11-101-9130 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9822 Supplier MilliporeSigma Supelco Supplier No. 030795ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethylbenzene is a colorless liquid, which has an aromatic odor. It is widely used in the manufacturing of styrene and as a solvent. It is insoluble in water.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 100-41-4
Ignition Point 810°F
Vapor Pressure 19 mmHg (37.7°C)
Vapor Density 3.7 (vs. air)
Density 0.867 g/mL (at 25°C)
Flash Point 15°C
Linear Formula C6H5C2H5
MDL Number MFCD00011647
Melting Point -95°C
Boiling Point 136°C
Synonym NSC 406903; Phenylethane
Type Analytical Standard
Quantity 5 mL
Formula Weight 106.17
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.