Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylbenzene solution NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm x 8 in. | 100-41-4 | MFCD00011647 | 1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 5513411EA
Encompass
Ethylbenzene solution NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm x 8 in. | Mol Wt: 106.17 | 100-41-4 | MFCD00011647 | 1EA
Catalog No. 50-180-6977