Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylbenzene solution NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), NMR tube size 8 mm x 7 in. | 100-41-4 | MFCD00011647 | 10X7IN
Catalog No. 501806979
Encompass
Ethylbenzene solution NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), NMR tube size 8 mm x 7 in. | Mol Wt: 106.17 | 100-41-4 | MFCD00011647 | 10X7IN
Catalog No. 50-180-6979 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 61245610X7IN
Ethylbenzene solution NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), NMR tube size 8 mm x 7 in. | Mol Wt: 106.17 | 100-41-4 | MFCD00011647 | 10X7IN