Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylbenzene solution NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm x 8 in. | 100-41-4 | MFCD00011647 | 10X8IN
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 61248010X8IN
Encompass
Ethylbenzene solution NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm x 8 in. | Mol Wt: 106.17 | 100-41-4 | MFCD00011647 | 10X8IN
Catalog No. 50-180-6980