Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylbenzene solution NMR reference standard, 0.4% in chloroform-d (99.8 atom %), NMR tube size 3 mm x 8 in. | 100-41-4 | MFCD00011647 | 10X8IN
Catalog No. 501806982
Encompass
Ethylbenzene solution NMR reference standard, 0.4% in chloroform-d (99.8 atom %), NMR tube size 3 mm x 8 in. | Mol Wt: 106.17 | 100-41-4 | MFCD00011647 | 10X8IN
Catalog No. 50-180-6982 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 61275810X8IN
Ethylbenzene solution NMR reference standard, 0.4% in chloroform-d (99.8 atom %), NMR tube size 3 mm x 8 in. | Mol Wt: 106.17 | 100-41-4 | MFCD00011647 | 10X8IN