Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylbenzene Solution, NMR Reference Standard, 0.1% in chloroform-d (99.8 atom% D), TMS 0.01%, MilliporeSigma™ Supelco™

Catalog No. 111018587 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
0.235 mL
0.7 mL
Packaging:
3 mm O.D. tube
5 mm O.D. x 8 in. tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-101-8587 0.7 mL 5 mm O.D. x 8 in. tube
11-101-8379 0.235 mL 3 mm O.D. tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8587 Supplier MilliporeSigma Supelco Supplier No. 4871048IN
Add to Cart
Add to Cart
Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging 5 mm O.D. x 8 in. tube
CAS 100-41-4
Linear Formula C8H10
MDL Number MFCD00011647
Type NMR Reference Standard
Quantity 0.7 mL
Formula Weight 106.17
Grade NMR Reference Standard
Concentration or Composition (by Analyte or Components) 0.1% in chloroform-d (99.8 atom % D)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.