Learn More
Medchemexpress LLC Ethylboronic acid | 4433-63-0 | 73.89 | 10 G
SDP
Catalog No. 5000447579
Encompass_Preferred
Ethylboronic acid is an organic boron compound used as a drug intermediate to construct macrocycles, capsules, polymers, and covalent organic frameworks (COFs).
  • Solid, white to yellow appearance.
  • Used to construct macrocycles, capsules, polymers, and covalent organic frameworks (COFs).
  • High purity of 99.87%.
Catalog No. 50-004-47579 Supplier Medchemexpress LLC Supplier No. HYW00093310G
Ethylboronic acid is an organic boron compound used as a drug intermediate to construct macrocycles, capsules, polymers, and covalent organic frameworks (COFs).
  • Solid, white to yellow appearance.
  • Used to construct macrocycles, capsules, polymers, and covalent organic frameworks (COFs).
  • High purity of 99.87%.