Learn More
Medchemexpress LLC Ethylboronic acid | 4433-63-0 | 73.89 | 5 G
SDP
Catalog No. 5000447578
Encompass_Preferred
Ethylboronic acid is an organic boron compound that serves as a drug intermediate for constructing macrocycles, capsules, polymers, and covalent organic frameworks (COFs). This product is for research use only.
  • Organic boron compound
  • Functions as a drug intermediate
  • Used to construct macrocycles, capsules, polymers, and covalent organic frameworks (COFs)
  • Soluble in DMSO at 200 mg/mL
Catalog No. 50-004-47578 Supplier Medchemexpress LLC Supplier No. HYW0009335G
Ethylboronic acid is an organic boron compound that serves as a drug intermediate for constructing macrocycles, capsules, polymers, and covalent organic frameworks (COFs). This product is for research use only.
  • Organic boron compound
  • Functions as a drug intermediate
  • Used to construct macrocycles, capsules, polymers, and covalent organic frameworks (COFs)
  • Soluble in DMSO at 200 mg/mL