Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethylboronic acid | 4433-63-0 | 73.89 | 50 G
SDP

Catalog No. 5000447117
Encompass_Preferred

Ethylboronic acid is an organic boron compound used as a drug intermediate. It serves in the construction of macrocycles, capsules, polymers, and covalent organic frameworks (COFs). It is intended for research use only.

  • Can be used for constructing macrocycles
  • Can be used for constructing capsules
  • Can be used for constructing polymers
  • Can be used for constructing covalent organic frameworks (COFs)
  • Appearance: white to yellow solid
  • Purity of 99.87%

Catalog No. 50-004-47117 Supplier Medchemexpress LLC Supplier No. HYW00093350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethylboronic acid is an organic boron compound used as a drug intermediate. It serves in the construction of macrocycles, capsules, polymers, and covalent organic frameworks (COFs). It is intended for research use only.

  • Can be used for constructing macrocycles
  • Can be used for constructing capsules
  • Can be used for constructing polymers
  • Can be used for constructing covalent organic frameworks (COFs)
  • Appearance: white to yellow solid
  • Purity of 99.87%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.