Learn More
Medchemexpress LLC Ethylboronic acid | 4433-63-0 | MFCD01074536 | 73.89 | 100 G
SDP
Catalog No. 5000447562
Encompass_Preferred
Ethylboronic acid is an organic boron compound used as a drug intermediate to construct macrocycles, capsules, polymers, and covalent organic frameworks (COFs).
  • Solid appearance
  • White to yellow color
  • Soluble in DMSO at 200 mg/mL (2706.73 mM)
  • Requires ultrasonic for solubility
  • Newly opened DMSO recommended due to hygroscopic nature
  • Classified as a biochemical assay reagent
  • Classified as a drug intermediate
Catalog No. 50-004-47562 Supplier Medchemexpress LLC Supplier No. HYW000933100G
Ethylboronic acid is an organic boron compound used as a drug intermediate to construct macrocycles, capsules, polymers, and covalent organic frameworks (COFs).
  • Solid appearance
  • White to yellow color
  • Soluble in DMSO at 200 mg/mL (2706.73 mM)
  • Requires ultrasonic for solubility
  • Newly opened DMSO recommended due to hygroscopic nature
  • Classified as a biochemical assay reagent
  • Classified as a drug intermediate