Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethylboronic acid

Catalog No. CDS003542 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS003542 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS003542 Supplier Sigma Aldrich Supplier No. CDS003542250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Heptafluoro-2-iodopropane is a secondary fluoro alkyl iodide. it can be prepared by addition of iodine monofluoride to hexafluoropropene.heptafluoro-2-iodopropane may be used in the preparation of perfluoropropyl magnesium bromide and perfluoropropyl magnesium iodide. it may be used as chain-transfer agent in telomerization reactions of:
  • 1,1-difluoroethylene and tetrafluoroethylene
  • Cf3i, c2f5i and n-c3f7i with 1,1-difluoroethylene
  • N-c3f7i with tetrafluoroethylene

Specifications

Molecular Formula C2H7BO2
Linear Formula C2H7BO2
Quantity 25 g
MDL Number MFCD01074536
Molecular Weight (g/mol) 73.89
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.