Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. E09135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0913-5G 5 g
E09131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0913-5G Supplier TCI America Supplier No. E09135G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4433-63-0
Molecular Formula C2H7BO2
Molecular Weight (g/mol) 73.89
MDL Number MFCD01074536
InChI Key PAVZHTXVORCEHP-UHFFFAOYSA-N
PubChem CID 521157
IUPAC Name ethylboronic acid
SMILES CCB(O)O

Specifications

Color White-Yellow
Quantity 5 g
Synonym boronic acid, ethyl, ethaneboronic acid, ethyldihydroxyborane, ethyl boronic acid, boronic acid, ethyl-9 ci
Formula Weight 73.89
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.