Learn More
Cayman Chemical EthylD-GlucuronIde 10mg
SDP
Catalog No. 502987824
Encompass_Preferred
A minor metabolite of ethanol; hair levels are positively correlated with alcohol consumption in patients with alcohol dependence syndrome
Catalog No. 50-298-7824 Supplier Cayman Chemical Supplier No. 2227110 MG
A minor metabolite of ethanol; hair levels are positively correlated with alcohol consumption in patients with alcohol dependence syndrome