Learn More
Cayman Chemical EthylD-GlucuronIde-d5 10mg
SDP
Catalog No. 502987835
Encompass_Preferred
An internal standard for the quantification of ethyl-β-D-glucuronide by GC- or LC-MS
Catalog No. 50-298-7835 Supplier Cayman Chemical Supplier No. 2227310 MG
An internal standard for the quantification of ethyl-β-D-glucuronide by GC- or LC-MS