Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle1-(3-aminobenzyl)piperidine-4-carboxylate, 97%, Thermo Scientific™

Catalog No. SPB08422DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPB08422DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB08422DA Supplier Thermo Scientific Chemicals Supplier No. SPB08422DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 306937-22-4
Molecular Formula C15H22N2O2
Molecular Weight (g/mol) 262.353
MDL Number MFCD02180894
InChI Key WXYBQSXOZUFNJY-UHFFFAOYSA-N
PubChem CID 2800723
IUPAC Name ethyl 1-[(3-aminophenyl)methyl]piperidine-4-carboxylate
SMILES CCOC(=O)C1CCN(CC1)CC2=CC(=CC=C2)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym ethyl 1-3-aminobenzyl piperidine-4-carboxylate, ethyl 1-3-aminophenyl methyl piperidine-4-carboxylate, 4-piperidinecarboxylicacid, 1-3-aminophenyl methyl-, ethyl ester, ethyl1-3-aminobenzyl piperidine-4-carboxylate, 1-3-aminobenzyl-4-ethoxycarbonyl piperidine, 3-4-ethoxycarbonyl piperidin-1-yl methyl aniline, 1-3-aminophenyl methyl-4-piperidinecarboxylic acid ethyl ester
Formula Weight 262.35
Percent Purity 97%
Chemical Name or Material ethyle1-(3-aminobenzyl)piperidine-4-carboxylate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.