Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle2-(phenylthio)acetate, 97%, Thermo Scientific™

Catalog No. KM08693EA
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KM08693EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM08693EA Supplier Thermo Scientific Chemicals Supplier No. KM08693EA
Add to Cart

Chemical Identifiers

CAS 7605-25-6
Molecular Formula C10H12O2S
Molecular Weight (g/mol) 196.264
MDL Number MFCD00145107
InChI Key SEDRTXNDGKRHBL-UHFFFAOYSA-N
Synonym ethyl phenylthio acetate, ethyl 2-phenylthio acetate, ethyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, ethyl ester, ethyl 2-phenylthioacetate, phenylthio acetic acid ethyl ester, phenylthio acetic acid, ethyl ester, ethyl phenylsulfanyl acetate, pubchem10853
PubChem CID 97312
IUPAC Name ethyl 2-phenylsulfanylacetate
SMILES CCOC(=O)CSC1=CC=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula C6H5SCH2C(O)OCH2CH3
Quantity 10 g
Formula Weight 196.27
Percent Purity 97%
Chemical Name or Material Ethyl 2-(phenylthio)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.