Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic anhydride, 99+%

Catalog No. AC147810025
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle

Trifluoroacetic anhydride, 99+%, Quantity: 500g, Packaging: Glass Bottle, Boiling Point: 39.1 deg.C, Undesignated, Melting Point: -63.5 deg.C, Molecular Weight: 210.03, Percent Purity: 99+%, Assay Percent Range: 99% min. (GC), Beilstein: 02, II, 186, CAS: 407-25-0 | CAS: 407-25-0 | C4F6O3 | 210.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 407-25-0
Molecular Formula C4F6O3
Molecular Weight (g/mol) 210.03
MDL Number MFCD00000416
InChI Key QAEDZJGFFMLHHQ-UHFFFAOYSA-N
Synonym trifluoroacetic anhydride, trifluoroacetic acid anhydride, 2,2,2-trifluoroacetic anhydride, bis trifluoroacetic anhydride, acetic acid, trifluoro-, anhydride, perfluoroacetic anhydride, trifluoroacetyl anhydride, tfaa, hexafluoroacetic anhydride, anhydrid kyseliny trifluoroctove
PubChem CID 9845
IUPAC Name (2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate
SMILES FC(F)(F)C(=O)OC(=O)C(F)(F)F

Specifications

Melting Point -63.5°C
Density 1.5100g/mL
Boiling Point 39.1°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.3
Linear Formula (CF3CO)2O
Quantity 2.5 kg
Beilstein 02, II, 186
Fieser 01,1221; 03,308; 05,701; 06,616; 07,389; 09,484; 12,530; 14,151; 15,14; 01,373
Specific Gravity 1.51
Solubility Information Solubility in water: hydrolysis. Other solubilities: miscible with polar organic solvents, partically soluble in unpolar hydrocarbons
Formula Weight 210.03
Vapor Pressure 141mbar at 20°C
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Trifluoroacetic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minu
EINECSNumber 206-982-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.