Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-oxocyclohexanecarboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AC379910010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC379910010 1 g Glass bottle
AC379910050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC379910010 Supplier Thermo Scientific Chemicals Supplier No. 379910010
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17159-79-4
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00013285
InChI Key ZXYAWONOWHSQRU-UHFFFAOYSA-N
Synonym ethyl 4-oxocyclohexanecarboxylate, ethyl 4-cyclohexanonecarboxylate, 4-ethoxycarbonyl cyclohexanone, ethyl cyclohexanone-4-carboxylate, ethyl-4-oxo-cyclohexane carboxylate, 4-oxo-cyclohexanecarboxylic acid ethyl ester, cyclohexanecarboxylic acid, 4-oxo-, ethyl ester, ethyl4-oxocyclohexanecarboxylate, 4-cyclohexanonecarboxylic acid ethyl ester, ethyl 4-oxocyclohexane carboxylate
PubChem CID 317638
IUPAC Name ethyl 4-oxocyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCC(=O)CC1

Specifications

Color Colorless to Yellow
Density 1.0760g/mL
Boiling Point 150°C to 152°C (40.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4580 to 1.461
Linear Formula (O=)C6H9CO2C2H5
Quantity 1 g
Specific Gravity 1.076
Solubility Information Solubility in water: insoluble
Formula Weight 170.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 4-oxocyclohexanecarboxylate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.