Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl benzoylacetate, 90%, Acros Organics

Click to view available options
Quantity:
5g
50g
100g
500g
Packaging:
Glass bottle
94-02-0
C11H12O3
192.21
MFCD00009196
GKKZMYDNDDMXSE-UHFFFAOYSA-N
ethyl benzoylacetate,benzoylacetic acid ethyl ester,ethyl benzoyl acetate,acetic acid, benzoyl-, ethyl ester,ethyl beta-oxobenzenepropanoate,ethylbenzoylacetate,benzoylacetic acid, ethyl ester,ethyl 3-phenyl-3-oxopropanoate,benzenepropanoic acid, beta-oxo-, ethyl ester,ethyl 3-oxo-3-phenylpropionate
7170
ethyl 3-oxo-3-phenylpropanoate
CCOC(=O)CC(=O)C1=CC=CC=C1
94-02-0
C11H12O3
192.21
MFCD00009196
GKKZMYDNDDMXSE-UHFFFAOYSA-N
ethyl benzoylacetate,benzoylacetic acid ethyl ester,ethyl benzoyl acetate,acetic acid, benzoyl-, ethyl ester,ethyl beta-oxobenzenepropanoate,ethylbenzoylacetate,benzoylacetic acid, ethyl ester,ethyl 3-phenyl-3-oxopropanoate,benzenepropanoic acid, beta-oxo-, ethyl ester,ethyl 3-oxo-3-phenylpropionate
7170
ethyl 3-oxo-3-phenylpropanoate
CCOC(=O)CC(=O)C1=CC=CC=C1
Boiling Point 265°C to 270°C
Flash Point 140°C
Specific Gravity 1.106
Refractive Index 1.5240 to 1.531
Color Brown
Density 1.1060g/mL
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Assay Percent Range 88% min. (HPLC)
Beilstein 10, 674
Merck Index 15, 3822
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols,ethers,miscible with oils
Formula Weight 192.21
Physical Form Powder
Percent Purity 90%
Chemical Name or Material Ethyl benzoylacetate
Show More Show Less

EINECSNumber : 202-295-3