Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl benzoylacetate, 90%

Catalog No. AC370850500
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC370850500 50 g Glass bottle
AC370850050 5 g Glass bottle
AC370851000 100 g Glass bottle
AC370855000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC370850500 Supplier Thermo Scientific Chemicals Supplier No. 370850500
Only null left
Add to Cart

CAS: 94-02-0 | C11H12O3 | 192.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-02-0
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
MDL Number MFCD00009196
InChI Key GKKZMYDNDDMXSE-UHFFFAOYSA-N
Synonym ethyl benzoylacetate, benzoylacetic acid ethyl ester, ethyl benzoyl acetate, acetic acid, benzoyl-, ethyl ester, ethyl beta-oxobenzenepropanoate, ethylbenzoylacetate, benzoylacetic acid, ethyl ester, ethyl 3-phenyl-3-oxopropanoate, benzenepropanoic acid, beta-oxo-, ethyl ester, ethyl 3-oxo-3-phenylpropionate
PubChem CID 7170
IUPAC Name ethyl 3-oxo-3-phenylpropanoate
SMILES CCOC(=O)CC(=O)C1=CC=CC=C1

Specifications

Color Brown
Density 1.1060g/mL
Boiling Point 265°C to 270°C
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.5240 to 1.531
Quantity 50 g
Beilstein 10, 674
Specific Gravity 1.106
Merck Index 15, 3822
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols,ethers,miscible with oils
Formula Weight 192.21
Percent Purity 90%
Physical Form Powder
Chemical Name or Material Ethyl benzoylacetate
Show More Show Less
EINECSNumber 202-295-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.