Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl isothiocyanate, 96%, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
542-85-8
C3H5NS
87.14
MFCD00004820
HBNYJWAFDZLWRS-UHFFFAOYSA-N
ethyl isothiocyanate,ethane, isothiocyanato,ethyl mustard oil,ethylisothiocyanate,isothiocyanic acid, ethyl ester,unii-3284mj2t8p,ccris 7323,ethylisothio-cyanate,isothiocyanato-ethane
10966
CHEBI:85098
isothiocyanatoethane
CCN=C=S
542-85-8
C3H5NS
87.14
MFCD00004820
HBNYJWAFDZLWRS-UHFFFAOYSA-N
ethyl isothiocyanate,ethane, isothiocyanato,ethyl mustard oil,ethylisothiocyanate,isothiocyanic acid, ethyl ester,unii-3284mj2t8p,ccris 7323,ethylisothio-cyanate,isothiocyanato-ethane
10966
CHEBI:85098
isothiocyanatoethane
CCN=C=S
Density 0.9950g/mL
Refractive Index 1.5100 to 1.513
Melting Point -6°C
Boiling Point 130°C to 132°C
Flash Point 32°C
Specific Gravity 0.995
Packaging Glass bottle
Quantity 5g
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Linear Formula C2H5NCS
Merck Index 15, 3870
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Formula Weight 87.14
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Ethyl isothiocyanate
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Causes serious eye damage.
Toxic in contact with skin.
Flammable liquid and vapor.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-831-2