Learn More
Cayman Chemical Ethylen Glycol mondodecyl100mg
SDP
Catalog No. 502909364
Encompass_Preferred
A nonionic surfactant; CMC = 27.5 µM at 25°C; enhances the transdermal permeation of the antiemetic ondansetron across isolated porcine skin at 160 mM, however, it decreases the viability of human epidermal keratinocytes and human dermal fibroblasts at 160 µM
Catalog No. 50-290-9364 Supplier Cayman Chemical Supplier No. 36857100 MG
A nonionic surfactant; CMC = 27.5 µM at 25°C; enhances the transdermal permeation of the antiemetic ondansetron across isolated porcine skin at 160 mM, however, it decreases the viability of human epidermal keratinocytes and human dermal fibroblasts at 160 µM