Learn More
Cayman Chemical Ethylen Glycol mondodecyl10mg
SDP
Catalog No. 502909754
Encompass_Preferred
A nonionic surfactant; CMC = 27.5 µM at 25°C; enhances the transdermal permeation of the antiemetic ondansetron across isolated porcine skin at 160 mM, however, it decreases the viability of human epidermal keratinocytes and human dermal fibroblasts at 160 µM
Catalog No. 50-290-9754 Supplier Cayman Chemical Supplier No. 3685710 MG
A nonionic surfactant; CMC = 27.5 µM at 25°C; enhances the transdermal permeation of the antiemetic ondansetron across isolated porcine skin at 160 mM, however, it decreases the viability of human epidermal keratinocytes and human dermal fibroblasts at 160 µM