Learn More
Cayman Chemical Ethylene Glycol monododecyl ether, Purity: ≥98%, CAS: 4536-30-5, 50 mg
SDP
Catalog No. 502909755
Encompass_Preferred
A nonionic surfactant; CMC = 27.5 µM at 25°C; enhances the transdermal permeation of the antiemetic ondansetron across isolated porcine skin at 160 mM, however, it decreases the viability of human epidermal keratinocytes and human dermal fibroblasts at 160 µM
Catalog No. 50-290-9755 Supplier Cayman Chemical Supplier No. 3685750 MG
A nonionic surfactant; CMC = 27.5 µM at 25°C; enhances the transdermal permeation of the antiemetic ondansetron across isolated porcine skin at 160 mM, however, it decreases the viability of human epidermal keratinocytes and human dermal fibroblasts at 160 µM