Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl N-piperazinecarboxylate, 99%

Catalog No. AC118872500
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC118872500 250 g
AC118870250 25 g
AC118871000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC118872500 Supplier Thermo Scientific Chemicals Supplier No. 118872500
Only null left
Add to Cart

CAS: 120-43-4 | C7H14N2O2 | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-43-4
Molecular Formula C7H14N2O2
Molecular Weight (g/mol) 158.2
MDL Number MFCD00005964
InChI Key LNOQURRKNJKKBU-UHFFFAOYSA-N
Synonym ethyl n-piperazinecarboxylate, ethyl 1-piperazinecarboxylate, n-carbethoxypiperazine, 1-carbethoxypiperazine, 1-carboethoxypiperazine, 1-piperazinecarboxylic acid, ethyl ester, n-ethoxycarbonyl piperazine, ethylcarbonyl piperazine, 1-ethoxycarbonyl piperazine, ethyl 1-piperazinocarboxylate
PubChem CID 8431
IUPAC Name ethyl piperazine-1-carboxylate
SMILES CCOC(=O)N1CCNCC1

Specifications

Color Yellow
Density 1.0800g/mL
Boiling Point 273°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4760 to 1.478
Quantity 250 g
Beilstein 23, II, 9
Specific Gravity 1.08
Solubility Information Solubility in water: freely soluble
Formula Weight 158.2
Percent Purity 99%
Physical Form Oily Liquid
Chemical Name or Material Ethyl N-piperazinecarboxylate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 204-395-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.