Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylene 99.9910G
Catalog No. 502907861
Encompass
Ethylene (ET) a simple olefin is an important synthetic reagent. It is also a plant hormone that regulates various physiological and developmental processes such as ripening of fruit abscission senescence and responses to wounding in plants. Pd(II) and Ni(II)-based catalysts are reported to catalyze the polymerization of ET to afford high molecular weight polymers. Mechanism of Diels-Alder Reaction of 1 3-butadiene with ET has been investigated. Diels-Alder reaction between butadiene and ET via concerted and the stepwise mechanisms has been investigated by ab initio MO methods. Dehydration of algae-produced ethanol to ET in the presence of nanoscale catalyst HZSM-5 (protonated Zeolite Socony Mobil-5) shows promising results in replacing fossil fuel methods for the industrial production of ET. The utility of microsized spent tea leaf powder (MSTLP) as an ethylene adsorbent has been investigated.
Catalog No. 50-290-7861 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 536164110G
Ethylene (ET) a simple olefin is an important synthetic reagent. It is also a plant hormone that regulates various physiological and developmental processes such as ripening of fruit abscission senescence and responses to wounding in plants. Pd(II) and Ni(II)-based catalysts are reported to catalyze the polymerization of ET to afford high molecular weight polymers. Mechanism of Diels-Alder Reaction of 1 3-butadiene with ET has been investigated. Diels-Alder reaction between butadiene and ET via concerted and the stepwise mechanisms has been investigated by ab initio MO methods. Dehydration of algae-produced ethanol to ET in the presence of nanoscale catalyst HZSM-5 (protonated Zeolite Socony Mobil-5) shows promising results in replacing fossil fuel methods for the industrial production of ET. The utility of microsized spent tea leaf powder (MSTLP) as an ethylene adsorbent has been investigated.