Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene in Air, 10 ppm, MilliporeSigma™ Supelco™

Catalog No. 111016474 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
14 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6474 14 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6474 Supplier MilliporeSigma Supelco Supplier No. 501379

Specifications:Contents: 14 liters Pressure: 240 psig Outlet fitting: CGA-160-1/8 inch, NPT F Weight: 1.5 lb.Dimensions: 3 x 11 inches D.O.T. specifications: 4B240

Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Packaging Cylinder
CAS 132259-10-0
Physical Form Gas
Type Analytical Standard
Quantity 14 L
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 10 ppm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.