Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Carbonate, +99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
96-49-1
C3H4O3
88.06
MFCD00005382
KMTRUDSVKNLOMY-UHFFFAOYSA-N
ethylene carbonate,glycol carbonate,ethylene glycol carbonate,dioxolone-2,cyclic ethylene carbonate,ethylene carbonic acid,1,3-dioxacyclopentan-2-one,2-dioxolone,carbonic acid, cyclic ethylene ester,texacar ec
7303
1,3-dioxolan-2-one
C1COC(=O)O1
96-49-1
C3H4O3
88.06
MFCD00005382
KMTRUDSVKNLOMY-UHFFFAOYSA-N
ethylene carbonate,glycol carbonate,ethylene glycol carbonate,dioxolone-2,cyclic ethylene carbonate,ethylene carbonic acid,1,3-dioxacyclopentan-2-one,2-dioxolone,carbonic acid, cyclic ethylene ester,texacar ec
7303
1,3-dioxolan-2-one
C1COC(=O)O1
Alkylation reagent
Viscosity 1.92 mPa.s (40°C)
Quantity 250g
Color White to Yellow
Flash Point 150°C
Specific Gravity 1.321
Density 1.3210g/mL
Boiling Point 246.7°C
Melting Point 34.0°C to 37.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99+%
Beilstein 19, 100
Fieser 01,372
Solubility Information Solubility in water: 214g/L (20°C). Other solubilities: soluble in ether,n-butanol and carbon tetra-,chloride,miscible with alcohol,ethyl acetate,benzene and,chloroform
Formula Weight 88.06
Physical Form Crystalline Low Melting Solid
Percent Purity ≥99%
Chemical Name or Material Ethylene carbonate
Show More Show Less

GHS H Statement:
Causes serious eye damage.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning