Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene chlorophosphate, 95%, Thermo Scientific Chemicals

Catalog No. aaa1832006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1832006 5 g
AAA1832014 25 g
AAA1832022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1832006 Supplier Thermo Scientific Chemicals Supplier No. A1832006
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene chlorophosphate is used in the syntheses of 2-methacryloyoxyethylphophocholine, phosphatidylcholines, 2′,3′-O-16-hentriacontanyliden-adenosine-5′-phosphocholine and hexadecylphosphocholine. It is also involved the synthesis of alpha-naphthylphosphorylcholine, which is used to detect phospholipase C activity. It is an important reactant for the synthesis of a zwitterionic silane, UV-polymerizable lipids utilizing Chabrier reaction and block copolymers of poly(aliphatic ester) clickable polyphosphoester.

Solubility
Miscible with water and chloroform.

Notes
Store in cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water and reacts violently.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6609-64-9
Molecular Formula C2H4ClO3P
Molecular Weight (g/mol) 142.475
MDL Number MFCD00043138
InChI Key SBMUNILHNJLMBF-UHFFFAOYSA-N
Synonym 2-chloro-2-oxo-1,3,2-dioxaphospholane, ethylene chlorophosphate, 2-chloro-1,3,2-dioxaphospholane-2-oxide, 2-chloro-1,3,2-dioxaphospholane 2-oxide, ethylene glycol chlorophosphate, soqdebhb^bsvusup@, 2-chloro-1,3,2??-dioxaphospholan-2-one, 2-chlor-1,3,2-dioxaphospholan-2-oxid, acmc-1b2u0, ethylene cycl-chlorophosphate
PubChem CID 81087
IUPAC Name 2-chloro-1,3,2$l^{5}-dioxaphospholane 2-oxide
SMILES C1COP(=O)(O1)Cl

Specifications

Melting Point 12°C to 14°C
Density 1.56
Boiling Point 98°C to 99°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.45
Quantity 5 g
UN Number UN3265
Beilstein 606582
Sensitivity Moisture sensitive
Solubility Information Miscible with water and chloroform.
Formula Weight 142.48
Percent Purity 95%
Chemical Name or Material Ethylene chlorophosphate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 229-560-6
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.